missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 2-Furoyl-Leu-Ile-Gly-Arg-Leu-Orn-NH_2
For cellular and molecular biology applications
3550.44 SEK
Specifications
Purification Method | HPLC |
---|---|
Common Name | Protease-Activated Receptor 2 (PAR2) Agonist |
Content And Storage | -30°C to -10°C |
Format | Lyophilized Powder |
Molecular Weight (g/mol) | 777.97 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
HPLC | |
-30°C to -10°C | |
777.97 | |
RUO - research use only |
Protease-Activated Receptor 2 (PAR2) Agonist | |
Lyophilized Powder | |
2-Furoyl-Leu-Ile-Gly-Arg-Leu-Orn-NH_2 | |
2-Furoyl-LIGRLO-NH2 |
For Research Use Only