Läs mer
2-[[(Guanidino)(imino)methyl]sulfanyl]ethanesulfonic Acid, TRC
Produktkod. 30010145
Change view
Kvantitet:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010145 500 mg
30012292 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30010145 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG835250500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1391053-66-9
Molekylformel C4 H10 N4 O3 S2
Molekylvikt (g/mol) 226.28
Synonym 2-[[[(Aminoiminomethyl)amino]iminomethyl]thio]ethanesulfonic acid, 2-[[(Guanidino)(imino)methyl]sulfanyl]ethanesulfonic Acid, Mesna Imp. B (EP)
IUPAC-namn 2-(N-carbamimidoylcarbamimidoyl)sulfanylethanesulfonic acid
LEDER NC(=N)NC(=N)SCCS(=O)(=O)O
Specifikationer
Kemiskt namn eller material 2-[((Guanidino)(imino)methyl)sulfanyl]ethanesulfonic Acid
Kvantitet 500 mg
InChI formel InChI=1S/C4H10N4O3S2/c5-3(6)8-4(7)12-1-2-13(9,10)11/h1-2H2,(H,9,10,11)(H5,5,6,7,8)
Formel vikt 226.0194