missing translation for 'onlineSavingsMsg'
Learn More

2-Hydrazino-2-imidazoline hydrobromide, 98%, Thermo Scientific Chemicals

Catalog Number p-7025066
Click to view available options
Quantity:
2.5 g
10 g
This item is not returnable. View return policy
55959-84-7
C3H9BrN4
181.037
MFCD00012694
HIJRJVVSABWXKG-UHFFFAOYSA-N
2-hydrazino-2-imidazoline hydrobromide, 2-hydrazinyl-4,5-dihydro-1h-imidazole hydrobromide, 2-hydrazino-delta-2-imidazoline hydrobromide, 4,5-dihydroimidazol-2-ylhydrazine hydrobromide, 4,5-dihydro-2-hydrazino-1h-imidazole hydrobromide, 2-hydrazino-4,5-dihydro-1h-imidazole hydrobromide, 4,5-dihydro-1h-imidazol-2-ylhydrazine hydrobromide, 4,5-dihydro-1h-imidazol-2-yl-hydrazine hydrobromide, 4,5-dihydro-1h-imidazol-2-yl-hydrazine, acmc-1av1k
3085099
4,5-dihydro-1H-imidazol-2-ylhydrazine;hydrobromide
C1CN=C(N1)NN.Br
This item is not returnable. View return policy
55959-84-7
C3H9BrN4
181.037
MFCD00012694
HIJRJVVSABWXKG-UHFFFAOYSA-N
2-hydrazino-2-imidazoline hydrobromide, 2-hydrazinyl-4,5-dihydro-1h-imidazole hydrobromide, 2-hydrazino-delta-2-imidazoline hydrobromide, 4,5-dihydroimidazol-2-ylhydrazine hydrobromide, 4,5-dihydro-2-hydrazino-1h-imidazole hydrobromide, 2-hydrazino-4,5-dihydro-1h-imidazole hydrobromide, 4,5-dihydro-1h-imidazol-2-ylhydrazine hydrobromide, 4,5-dihydro-1h-imidazol-2-yl-hydrazine hydrobromide, 4,5-dihydro-1h-imidazol-2-yl-hydrazine, acmc-1av1k
3085099
4,5-dihydro-1H-imidazol-2-ylhydrazine;hydrobromide
C1CN=C(N1)NN.Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 181°C to 185°C
Quantity 2.5 g
Sensitivity Hygroscopic
Formula Weight 181.04
Percent Purity 98%
Chemical Name or Material 2-Hydrazino-2-imidazoline hydrobromide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'