missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxy-1,4-naphthoquinone, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
2-Hydroxy-1,4-naphthoquinone is used for preparing decorative hair and skin dyes. 2-Hydroxy-1,4-naphthoquinone also demonstrates antimicrobial and antioxidant effects. It also suppress the formation of hydrogen peroxide and superoxide radical anion by aldehyde oxidase-catalyzed reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Hydroxy-1,4-naphthoquinone is used for preparing decorative hair and skin dyes. 2-Hydroxy-1,4-naphthoquinone also demonstrates antimicrobial and antioxidant effects. It also suppress the formation of hydrogen peroxide and superoxide radical anion by aldehyde oxidase-catalyzed reactions.
Solubility
Soluble in water. 2 g/L at 20°C
Notes
Incompatible with strong oxidizing agents. Store at room temperature.
2-Hydroxy-1,4-naphthoquinone is used for preparing decorative hair and skin dyes. 2-Hydroxy-1,4-naphthoquinone also demonstrates antimicrobial and antioxidant effects. It also suppress the formation of hydrogen peroxide and superoxide radical anion by aldehyde oxidase-catalyzed reactions.
Solubility
Soluble in water. 2 g/L at 20°C
Notes
Incompatible with strong oxidizing agents. Store at room temperature.
Specifications
Melting Point | ∼193°C (decomposition) |
Quantity | 10 g |
Beilstein | 1565260 |
Merck Index | 14,5393 |
Solubility Information | Soluble in water. 2g/L at 20°C |
Formula Weight | 174.16 |
Percent Purity | ≥98% |
Chemical Name or Material | 2-Hydroxy-1,4-naphthoquinone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'