Läs mer
2-Hydroxy-1-phenyl-1-propanone, TRC
Produktkod. 30007860
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007860 250 mg
30009991 500 mg
30003303 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007860 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH950250250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5650-40-8
Molekylformel C9 H10 O2
Molekylvikt (g/mol) 150.17
Synonym 1-Propanone, 2-hydroxy-1-phenyl-, Propiophenone, 2-hydroxy- (7CI,8CI), Propiophenone, α-hydroxy- (3CI), 2-Hydroxy-1-phenyl-1-propanone, 1-Hydroxyethyl phenyl ketone, 1-Phenyl-2-hydroxy-1-propanone, 2-Hydroxypropiophenone, Benzoylmethylcarbinol, Lactophenone, Methanol, benzoylmethyl-, NSC 402216, α-Hydroxypropiophenone
IUPAC-namn 2-hydroxy-1-phenylpropan-1-one
LEDER CC(O)C(=O)c1ccccc1
Specifikationer
Kemiskt namn eller material 2-Hydroxy-1-phenyl-1-propanone
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C9H10O2/c1-7(10)9(11)8-5-3-2-4-6-8/h2-7,10H,1H3
Formel vikt 150.0681
Procent renhet >95%