missing translation for 'onlineSavingsMsg'
Läs mer

2-Hydroxy-1-phenylpentane, TRC

Produktkod. 30405943
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30405943 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30405943 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH95089550MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 2-Hydroxy-1-phenylpentane
CAS 705-73-7
Molekylformel C11H16O
Kvantitet 50 mg
Synonym alpha-Propylphenethyl Alcohol, (±)-1-Phenyl-2-pentanol, 1-Phenyl-2-pentanol, alpha-Propylbenzeneethanol
InChI formel InChI=1S/C11H16O/c1-2-6-11(12)9-10-7-4-3-5-8-10/h3-5,7-8,11-12H,2,6,9H2,1H3
LEDER CCCC(O)Cc1ccccc1
IUPAC-namn 1-phenylpentan-2-ol
Molekylvikt (g/mol) 164.24
Formel vikt 164.1201
Produkttitel
Välj ett problem

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.