Läs mer
2-Hydroxy-3-metylsmörsyra, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | 2-Hydroxy-3-methylbutyric Acid |
| CAS | 4026-18-0 |
| Molekylformel | C5 H10 O3 |
| Kvantitet | 1 g |
| Synonym | Butanoic acid, 2-hydroxy-3-methyl-, Butyric acid, 2-hydroxy-3-methyl- (6CI,7CI,8CI), 2-Hydroxy-3-methylbutanoic acid, (±)-2-Hydroxy-3-methylbutyric acid, (±)-2-Hydroxyisopentanoic acid, (±)-2-Hydroxyisovaleric acid, (±)-α-Hydroxyisovaleric acid, 2-Hydroxy-3-methylbutyric acid, 2-Hydroxyisovaleric acid, 2-Oxyisovaleric acid, 3-Methyl-2-hydroxybutyric acid, DL-2-Hydroxy-3-methylbutanoic acid, DL-2-Hydroxyisovaleric acid, DL-α-Hydroxyisovaleric acid, NSC 227884, dl-2-Hydroxyisovaleric acid, α-Hydroxyisovaleric acid, α-Hydroxyl-3-methylbutyric acid |
| InChI formel | InChI=1S/C5H10O3/c1-3(2)4(6)5(7)8/h3-4,6H,1-2H3,(H,7,8) |
| LEDER | CC(C)C(O)C(=O)O |
| IUPAC-namn | 2-hydroxy-3-metylbutansyra |
| Molekylvikt (g/mol) | 118.13 |
| Formel vikt | 118.063 |