Läs mer
2-Hydroxy-3-phenylpyrazine, TRC
Produktkod. 30136762
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136762 100 mg
30134776 25 mg
30005799 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136762 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH950200100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2882-18-0
Molekylformel C10 H8 N2 O
Molekylvikt (g/mol) 172.18
Synonym 3-Phenyl-2(1H)-pyrazinone, Pyrazinol, 3-phenyl- (6CI,7CI,8CI), 2-Hydroxy-3-phenylpyrazine, 3-Phenyl-1H-pyrazin-2-one, 3-Phenyl-2-pyrazinol
IUPAC-namn 3-phenyl-1H-pyrazin-2-one
LEDER O=C1NC=CN=C1c2ccccc2
Specifikationer
Kemiskt namn eller material 2-Hydroxy-3-phenylpyrazine
Kvantitet 100 mg
InChI formel InChI=1S/C10H8N2O/c13-10-9(11-6-7-12-10)8-4-2-1-3-5-8/h1-7H,(H,12,13)
Formel vikt 172.0637