Läs mer
2-Hydroxy-4-amino Butanoic Acid, TRC
Produktkod. 30391604
Change view
Kvantitet:
25 g
Produktkod. Kvantitet
30391604 25 g
1 options
Produktkod. 30391604 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH79729325G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Hydroxy-4-amino Butanoic Acid
CAS 13477-53-7
Molekylformel C4H9NO3
Kvantitet 25 g
Synonym 4-Amino-2-hydroxybutanoic Acid, (RS)-2-Hydroxy-GABA, (RS)-2-Hydroxy-gamma-aminobutyric Acid, 2-Hydroxy-gamma-aminobutyric Acid, 4-Amino-2-hydroxybutyric Acid, DL-4-Amino-2-hydroxybutanoic Acid, DL-4-Amino-2-hydroxybutyric Acid, HABAgamma-Amino-alpha-hydroxybutyric Acid
InChI formel InChI=1S/C4H9NO3/c5-2-1-3(6)4(7)8/h3,6H,1-2,5H2,(H,7,8)
LEDER NCCC(O)C(=O)O
IUPAC-namn 4-amino-2-hydroxybutanoic acid
Molekylvikt (g/mol) 119.12
Formel vikt 119.0582