missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-methoxybenzophenone, 98%, Thermo Scientific Chemicals

Catalog Number p-145335
Click to view available options
Quantity:
5 g
100 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
131-57-7
C14H12O3
228.25
MFCD00008387
DXGLGDHPHMLXJC-UHFFFAOYSA-N
oxybenzone, 2-hydroxy-4-methoxybenzophenone, benzophenone-3, oxybenzon, 4-methoxy-2-hydroxybenzophenone, 2-benzoyl-5-methoxyphenol, oxibenzona, oxybenzonum, 2-hydroxy-4-methoxyphenyl phenyl methanone, anuvex
4632
CHEBI:34283
(2-hydroxy-4-methoxyphenyl)-phenylmethanone
COC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O
This item is not returnable. View return policy
131-57-7
C14H12O3
228.25
MFCD00008387
DXGLGDHPHMLXJC-UHFFFAOYSA-N
oxybenzone, 2-hydroxy-4-methoxybenzophenone, benzophenone-3, oxybenzon, 4-methoxy-2-hydroxybenzophenone, 2-benzoyl-5-methoxyphenol, oxibenzona, oxybenzonum, 2-hydroxy-4-methoxyphenyl phenyl methanone, anuvex
4632
CHEBI:34283
(2-hydroxy-4-methoxyphenyl)-phenylmethanone
COC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point >62.5°C
Color White to Yellow
Boiling Point 150°C to 160°C (5.0 mmHg)
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula HOC6H3(OCH3)COC6H5
Quantity 5 g
Beilstein 08,312
Merck Index 15,7053
Solubility Information Solubility in water: .. Other solubilities: readily soluble in most organic solvents
Formula Weight 228.25
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-4-methoxybenzophenone, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'