Läs mer
2-Hydroxy-alpha,alpha,4-trimethylcyclohexanemethanol, TRC
Produktkod. 30007209
Change view
Kvantitet:
25 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007209 500 mg
17933875 100 mg
17974125 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007209 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH972160500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 42822-86-6
Molekylformel C10 H20 O2
Molekylvikt (g/mol) 172.26
Synonym p-Menthane-3,8-diol (7CI), 2-Hydroxy-α,α,4-trimethylcyclohexanemethanol, 1-Hydroxy-2-(1-methyl-1-hydroxyethyl)-5-methylcyclohexane, 1-Hydroxy-5-methyl-2-(1-methyl-1-hydroxyethyl)cyclohexane, 3,8-p-Menthanediol, Citrepel, Citriodiol, Cooling agent 38D, Geranium cyclohexane, Geranodyle, p-Menthan-3,8-diol
IUPAC-namn 2-(2-hydroxypropan-2-yl)-5-methylcyclohexan-1-ol
LEDER CC1CCC(C(O)C1)C(C)(C)O
Specifikationer
Kemiskt namn eller material 2-Hydroxy-a,a,4-trimethylcyclohexanemethanol
Kvantitet 500 mg
InChI formel InChI=1S/C10H20O2/c1-7-4-5-8(9(11)6-7)10(2,3)12/h7-9,11-12H,4-6H2,1-3H3
Formel vikt 172.1463