Läs mer
2-Hydroxy Atorvastatin Dihydrate Monosodium Salt, TRC
Produktkod. 30008296
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008296 1 mg
30012555 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30008296 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH8289501MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1421760-64-6
Molekylformel C33H38FN2NaO8
Molekylvikt (g/mol) 632.65
Synonym (βR,δR)-2-(4-Fluorophenyl)-β,δ-dihydroxy-4-[[(2-hydroxyphenyl)amino]carbonyl]-5-(1-methylethyl)-3-phenyl-1H-pyrrole-1-heptanoic Acid Dihydrate Monosodium Salt, BMS 243887-01, PD 152873, o-Hydroxyatorvastatin Dihydrate Monosodium Salt;
LEDER O=C(C(C(C1=CC=CC=C1)=C(C2=CC=C(F)C=C2)N3CC[C@@H](O)C[C@@H](O)CC(O[Na])=O)=C3C(C)C)NC(C=CC=C4)=C4O
Specifikationer
Kemiskt namn eller material 2-Hydroxy Atorvastatin Dihydrate Monosodium Salt
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C33H35FN2O6.Na/c1-20(2)31-30(33(42)35-26-10-6-7-11-27(26)39)29(21-8-4-3-5-9-21)32(22-12-14-23(34)15-13-22)36(31)17-16-24(37)18-25(38)19-28(40)41;/h3-15,20,24-25,37-39H,16-19H2,1-2H3,(H,35,42)(H,40,41);/q;+1/p-1/t24-,25-;/m1./s1
Formel vikt 632.251
Procent renhet >95%