Läs mer
2-Hydroxy Estrone, TRC
Produktkod. 18012020
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18012020 1 mg
18003510 5 mg
17980735 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18012020 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9419451MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 362-06-1
Molekylformel C18 H22 O3
Molekylvikt (g/mol) 286.37
Synonym Estra-1,3,5(10)-trien-17-one, 2,3-dihydroxy-, 2,3-Dihydroxyestra-1,3,5(10)-trien-17-one, 2-Hydroxyestrone, Catecholestrone
IUPAC-namn (8R,9S,13S,14S)-2,3-dihydroxy-13-methyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-one
LEDER C[C@]12CC[C@H]3[C@@H](CCc4cc(O)c(O)cc34)[C@@H]1CCC2=O
Specifikationer
Kemiskt namn eller material 2-Hydroxy Estrone
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C18H22O3/c1-18-7-6-11-12(14(18)4-5-17(18)21)3-2-10-8-15(19)16(20)9-13(10)11/h8-9,11-12,14,19-20H,2-7H2,1H3/t11-,12+,14-,18-/m0/s1
Formel vikt 286.1569
Procent renhet >95%