Läs mer
2-Hydroxy Felbamate, TRC
Produktkod. 30010585
Change view
Kvantitet:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010585 25 mg
30003703 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30010585 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH94240525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 109482-32-8
Molekylformel C11H14N2O5
Molekylvikt (g/mol) 254.24
Synonym 2-Phenyl-1,2,3-propanetriol 1,3-Dicarbamate, 2-Hydroxy-2-phenyl-1,3-propanediyl dicarbamate, W 2292, W 2992;
IUPAC-namn (3-carbamoyloxy-2-hydroxy-2-phenylpropyl) carbamate
LEDER NC(=O)OCC(O)(COC(=O)N)c1ccccc1
Specifikationer
Kemiskt namn eller material 2-Hydroxy Felbamate
Kvantitet 25 mg
InChI formel InChI=1S/C11H14N2O5/c12-9(14)17-6-11(16,7-18-10(13)15)8-4-2-1-3-5-8/h1-5,16H,6-7H2,(H2,12,14)(H2,13,15)
Formel vikt 254.0903