Läs mer
2-Hydroxy Imipramine Beta-D-Glucuronide, TRC
Produktkod. 30404155
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30404155 1 mg
1 options
Produktkod. 30404155 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9434101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Hydroxy Imipramine beta-D-Glucuronide
CAS 54190-76-0
Molekylformel C25 H32 N2 O7
Kvantitet 1 mg
Synonym 2-Hydroxy Imipramine beta-D-Glucuronide
InChI formel InChI=1S/C25H32N2O7/c1-26(2)12-5-13-27-18-7-4-3-6-15(18)8-9-16-14-17(10-11-19(16)27)33-25-22(30)20(28)21(29)23(34-25)24(31)32/h3-4,6-7,10-11,14,20-23,25,28-30H,5,8-9,12-13H2,1-2H3,(H,31,32)/t20-,21-,22+,23-,25?/m0/s1
LEDER CN(C)CCCN1c2ccccc2CCc3cc(OC4O[C@@H]([C@@H](O)[C@H](O)[C@H]4O)C(=O)O)ccc13
IUPAC-namn (2S,3S,4S,5R)-6-[[11-[3-(dimethylamino)propyl]-5,6-dihydrobenzo[b][1]benzazepin-3-yl]oxy]-3,4,5-trihydroxyoxane-2-carboxylic acid
Molekylvikt (g/mol) 472.53
Formel vikt 472.221