Läs mer
2-Hydroxy-N-[(2R)-2-hydroxy-2-(3-hydroxyphenyl)ethyl]-N-methylacetamide, Mikromol™
Produktkod. 17922836
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17922836 100 mg
17901776 25 mg
30462165 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17922836 Leverantör Mikromol Leverantörsnummer MM0081.13
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-Hydroxy-N-[(2R)-2-hydroxy-2-(3-hydroxyphenyl)ethyl]-N-methylacetamide
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Hydroxy-N-[(2R)-2-hydroxy-2-(3-hydroxyphenyl)ethyl]-N-methylacetamide
CAS 2380769-62-8
Analyt- eller komponentnamn 2-Hydroxy-N-[(2R)-2-hydroxy-2-(3-hydroxyphenyl)ethyl]-N-methylacetamide
Frakt skick Room Temperature
Molekylformel C11 H15 N O4
Certifikat och efterlevnad ISO 9001
Synonym 2-Hydroxy-N-((2R)-2-hydroxy-2-(3-hydroxyphenyl)ethyl)-N-methylacetamide
InChI formel InChI=1S/C11H15NO4/c1-12(11(16)7-13)6-10(15)8-3-2-4-9(14)5-8/h2-5,10,13-15H,6-7H2,1H3/t10-/m0/s1
LEDER CN(C[C@H](O)c1cccc(O)c1)C(=O)CO
IUPAC-namn 2-hydroxy-N-[(2R)-2-hydroxy-2-(3-hydroxyphenyl)ethyl]-N-methylacetamide
Molekylvikt (g/mol) 225.24
Kvantitet 100 mg
Formel vikt 225.1001 g/mol