missing translation for 'onlineSavingsMsg'
Läs mer

2-Hydroxybenzaldehyde, Sigma-Aldrich™

Produktkod. 15888350
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mL
250 mL
1 L
Förpackningsstorlek:
1 L
250ml
5ml
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
15888350 1 L 1 L
15868350 5 mL 5ml
15878350 250 mL 250ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15888350 Supplier Sigma Aldrich Supplier No. 8006401000
 Begränsad artikel
Beräknad leverans: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-02-8
Kvalitet Research
Molekylvikt (g/mol) 122.12

Specifications

Smältpunkt -7°C
Densitet 1.17 g/cm3 at 20°C
MDL-nummer MFCD00003317
Kvantitet 1 L
Synonym 2-Hydroxybenzaldehyde, Salicylaldehyde, 2-Hydroxybenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.