missing translation for 'onlineSavingsMsg'
Läs mer

2-Hydroxybutanoic Acid, TRC

Product Code. 30012453
Change view
Click to view available options
Kvantitet:
2.5 g
10 g
25 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet unitSize
30012453 25 g -
30007315 10 g -
30003352 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012453 Supplier Toronto Research Chemicals Supplier No. TRCH83118525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 600-15-7
Molekylformel C4H8O3
Molekylvikt (g/mol) 104.1
Synonym (±)-2-Hydroxybutanoic Acid, DL-2-Hydroxybutyric Acid, (RS)-2-Hydroxybutyric Acid, (±)-2-Hydroxy-n-butyric Acid, (±)-2-Hydroxybutanoic Acid, (±)-2-Hydroxybutyric Acid, (±)-α-Hydroxybutyric Acid, 2-Hydroxybutanoic Acid, 2-Hydroxybutyric Acid, DL-2-Hydroxybutanoic Acid, DL-2-Hydroxybutyric Acid, DL-α-Hydroxybutyric Acid, NSC 6495, α-Hydroxy-n-butyric Acid, α-Hydroxybutanoic Acid, α-Hydroxybutyric Acid
IUPAC-namn 2-hydroxybutanoic acid
LEDER CCC(O)C(=O)O

Specifications

Kemiskt namn eller material 2-Hydroxybutanoic Acid
Kvantitet 25 g
Formel vikt 104.0473
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.