Läs mer
2-Hydroxyethyl Benzoate(Ethylene Glycol Monobenzoate), TRC
Produktkod. 30377134
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30377134 10 g
30373794 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30377134 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE91752510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 94-33-7
Molekylformel C9 H10 O3
Molekylvikt (g/mol) 166.17
Synonym 2-Benzoyloxyethanol, 1,2-Ethanediol, 1-benzoate, 1,2-Ethanediol, monobenzoate (9CI), Ethylene glycol, monobenzoate (8CI), 2-(Benzoyloxy)ethanol, 2-Benzoyloxy-1-hydroxyethane, 2-Hydroxyethyl benzoate, 2-Hydroxyethyl phenylacetate, NSC 57378, β-Hydroxyethyl benzoate
IUPAC-namn 2-hydroxyethyl benzoate
LEDER OCCOC(=O)c1ccccc1
Specifikationer
Kemiskt namn eller material 2-Hydroxyethyl Benzoate
Kvantitet 10 g
InChI formel InChI=1S/C9H10O3/c10-6-7-12-9(11)8-4-2-1-3-5-8/h1-5,10H,6-7H2
Formel vikt 166.063