Läs mer
2-Hydroxyethyl Methyl Terephthalate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | 2-Hydroxyethyl Methyl Terephthalate |
| CAS | 3645-00-9 |
| Molekylformel | C11 H12 O5 |
| Kvantitet | 500 mg |
| Synonym | 1,4-Benzenedicarboxylic Acid 2-Hydroxyethyl Methyl Ester, Terephthalic Acid 2-Hydroxyethyl Methyl Ester, Ethylene Glycol Mono(methyl (Terephthalate), 2-Hydroxyethyl Methyl Terephthalate, Dimethyl Terephthalate-Ethylene Glycol Monoester, Methyl 2-Hydroxyethyl Terephthalate, Monoglycol Mnnomethyl Terephthalate, β-Hydroxyethyl Methyl Terephthalate |
| InChI formel | InChI=1S/C11H12O5/c1-15-10(13)8-2-4-9(5-3-8)11(14)16-7-6-12/h2-5,12H,6-7H2,1H3 |
| LEDER | COC(=O)c1ccc(cc1)C(=O)OCCO |
| IUPAC-namn | 1-O-(2-hydroxyethyl) 4-O-methyl benzene-1,4-dicarboxylate |
| Molekylvikt (g/mol) | 224.21 |
| Formel vikt | 224.0685 |