Läs mer
2-Hydroxyethyl Nortadalafil, TRC
Produktkod. 30138014
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138014 100 mg
30135740 10 mg
30007366 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138014 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH942245100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 385769-94-8
Molekylformel C23H21N3O5
Molekylvikt (g/mol) 419.43
Synonym 2-Hydroxyethyl Nortadalafil, rel-(6R,12aR)-6-(1,3-Benzodioxol-5-yl)-2,3,6,7,12,12a-hexahydro-2-(2-hydroxyethyl)pyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione
IUPAC-namn (2R,8R)-2-(1,3-benzodioxol-5-yl)-6-(2-hydroxyethyl)-3,6,17-triazatetracyclo[8.7.0.0^{3,8}.0^{11,16}]heptadeca-1(10),11,13,15-tetraene-4,7-dione
LEDER OCCN1CC(=O)N2[C@H](Cc3c([nH]c4ccccc34)[C@H]2c5ccc6OCOc6c5)C1=O
Specifikationer
Kemiskt namn eller material 2-Hydroxyethyl Nortadalafil
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C23H21N3O5/c27-8-7-25-11-20(28)26-17(23(25)29)10-15-14-3-1-2-4-16(14)24-21(15)22(26)13-5-6-18-19(9-13)31-12-30-18/h1-6,9,17,22,24,27H,7-8,10-12H2/t17-,22-/m1/s1
Formel vikt 419.1481
Procent renhet >95%