missing translation for 'onlineSavingsMsg'
Läs mer

2-(Hydroxyimino)-2-(methylthio)acetic Acid Methyl Ester, TRC

Produktkod. 30403678
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30403678 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Product Code. 30403678 Supplier Toronto Research Chemicals Supplier No. TRCH9432901G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 2-(Hydroxyimino)-2-(methylthio)acetic Acid Methyl Ester
CAS 23135-20-8
Molekylformel C4H7NO3S
Kvantitet 1 g
Synonym Thio-oxalic Acid O,S-Dimethyl Ester 1-Oxime
InChI formel InChI=1S/C4H7NO3S/c1-8-4(6)3(5-7)9-2/h7H,1-2H3
LEDER COC(=O)C(=NO)SC
IUPAC-namn methyl 2-hydroxyimino-2-methylsulfanylacetate
Molekylvikt (g/mol) 149.17
Formel vikt 149.0147
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.