Läs mer
2-(Hydroxymethyl)-6-oxo-1,6-dihydro-[3,4'-bipyridine]-5-carbonitrile Trifluoroacetate Salt, TRC
Produktkod. 30407571
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30407571 100 mg
1 options
Produktkod. 30407571 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH825955100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(Hydroxymethyl)-6-oxo-1,6-dihydro-[3,4'-bipyridine]-5-carbonitrile Trifluoroacetate Salt
CAS 400063-09-4
Molekylformel C12H9N3O2 C2HF3O2
Kvantitet 100 mg
Synonym [3,4'-Bipyridine]-5-carbonitrile, 1,6-dihydro-2-(hydroxymethyl)-6-oxo-, 2,2,2-trifluoroacetate (1:1) (ACI), [3,4'-Bipyridine]-5-carbonitrile, 1,6-dihydro-2-(hydroxymethyl)-6-oxo-, mono(trifluoroacetate) (salt) (9CI), 1,6-Dihydro-2-(hydroxymethyl)-6-oxo[3,4'-bipyridine]-5-carbonitrile trifluoroacetate, 3-Cyano-6-hydroxymethyl-5-(4-pyridyl)-2-pyridone trifluoroacetate, 2-(Hydroxymethyl)-6-oxo-1,6-dihydro-[3,4'-bipyridine]-5-carbonitrile trifluoroacetate
InChI formel InChI=1S/C12H9N3O2.C2HF3O2/c13-6-9-5-10(8-1-3-14-4-2-8)11(7-16)15-12(9)17;3-2(4,5)1(6)7/h1-5,16H,7H2,(H,15,17);(H,6,7)
LEDER OCC1=C(C=C(C#N)C(=O)N1)c2ccncc2.OC(=O)C(F)(F)F
IUPAC-namn 6-(hydroxymethyl)-2-oxo-5-pyridin-4-yl-1H-pyridine-3-carbonitrile;2,2,2-trifluoroacetic acid
Molekylvikt (g/mol) 341.24
Formel vikt 341.0623