Läs mer
2-Hydroxymethyl Olanzapine, TRC
Produktkod. 30135431
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135431 5 mg
30134140 10 mg
30008776 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135431 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9477805MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 174756-45-7
Molekylformel C17 H20 N4 O S
Molekylvikt (g/mol) 328.43
Synonym 10H-Thieno[2,3-b][1,5]benzodiazepine-2-methanol, 4-(4-methyl-1-piperazinyl)-, 4-(4-Methyl-1-piperazinyl)-10H-thieno[2,3-b][1,5]benzodiazepine-2-methanol, LY 290411
IUPAC-namn [4-(4-methylpiperazin-1-yl)-10H-thieno[2,3-b][1,5]benzodiazepin-2-yl]methanol
LEDER CN1CCN(CC1)C2=Nc3ccccc3Nc4sc(CO)cc24
Specifikationer
Kemiskt namn eller material 2-Hydroxymethyl Olanzapine
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C17H20N4OS/c1-20-6-8-21(9-7-20)16-13-10-12(11-22)23-17(13)19-15-5-3-2-4-14(15)18-16/h2-5,10,19,22H,6-9,11H2,1H3
Formel vikt 328.1358
Procent renhet >95%