Läs mer
2-Hydroxypropyl Nortadalafil (Mixture of Diastereomers), TRC
Produktkod. 30136579
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136579 1 g
30004974 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136579 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9527051G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1353020-85-5
Molekylformel C24 H23 N3 O5
Molekylvikt (g/mol) 433.46
Synonym Pyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione, 6-(1,3-benzodioxol-5-yl)-2,3,6,7,12,12a-hexahydro-2-(2-hydroxypropyl)-, (6R,12aR)-, (6R,12aR)-6-(1,3-Benzodioxol-5-yl)-2,3,6,7,12,12a-hexahydro-2-(2-hydroxypropyl)pyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione, 2-Hydroxypropylnortadalafil, 2-Hydroxypropyl Nortadalafil
IUPAC-namn (2R,8R)-2-(1,3-benzodioxol-5-yl)-6-(2-hydroxypropyl)-3,6,17-triazatetracyclo[8.7.0.0^{3,8}.0^{11,16}]heptadeca-1(10),11,13,15-tetraene-4,7-dione
LEDER CC(O)CN1CC(=O)N2[C@H](Cc3c([nH]c4ccccc34)[C@H]2c5ccc6OCOc6c5)C1=O
Specifikationer
Kemiskt namn eller material 2-Hydroxypropyl Nortadalafil (mixture of diastereomers)
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C24H23N3O5/c1-13(28)10-26-11-21(29)27-18(24(26)30)9-16-15-4-2-3-5-17(15)25-22(16)23(27)14-6-7-19-20(8-14)32-12-31-19/h2-8,13,18,23,25,28H,9-12H2,1H3/t13?,18-,23-/m1/s1
Formel vikt 433.1638
Procent renhet >95%