Learn More
2-Hydroxypyridine-N-oxide, 97%, Thermo Scientific Chemicals
Catalog Number p-7077213
Click to view available options
Quantity:
25 g
100 g
500 g
13161-30-3
C5H5NO2
111.1
SNUSZUYTMHKCPM-UHFFFAOYSA-N
2-pyridinol-1-oxide, 2-hydroxypyridine 1-oxide, 2-hydroxypyridine n-oxide, 1-hydroxypyridin-2 1h-one, 2-hydroxypyridine-n-oxide, hydroxypyridinone, pyridinol, 1-oxide, 2-pyridinol 1-oxide, 1-hydroxy-2-pyridone, 1-hydroxy-1,2-dihydropyridin-2-one
69975
1-hydroxypyridin-2-one
C1=CC(=O)N(C=C1)O
13161-30-3
C5H5NO2
111.1
SNUSZUYTMHKCPM-UHFFFAOYSA-N
2-pyridinol-1-oxide, 2-hydroxypyridine 1-oxide, 2-hydroxypyridine n-oxide, 1-hydroxypyridin-2 1h-one, 2-hydroxypyridine-n-oxide, hydroxypyridinone, pyridinol, 1-oxide, 2-pyridinol 1-oxide, 1-hydroxy-2-pyridone, 1-hydroxy-1,2-dihydropyridin-2-one
69975
1-hydroxypyridin-2-one
C1=CC(=O)N(C=C1)O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 147.0°C to 152.0°C
Flash Point 100°C
Infrared Spectrum Authentic
Quantity 25 g
Formula Weight 111.1
Percent Purity 97%
Chemical Name or Material 2-Hydroxypyridine-N-oxide
compliance-icons
Product Identifier
  • 2-Hydroxypyridine-N-oxide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.