Learn More
2-Indanylacetic acid, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
2-Indanylacetic acid is the starting material in the synthesis of pentacylic core of (+)-Salvileucalin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Indanylacetic acid is the starting material in the synthesis of pentacylic core of (+)-Salvileucalin.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
2-Indanylacetic acid is the starting material in the synthesis of pentacylic core of (+)-Salvileucalin.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Melting Point | 91°C to 93°C |
Quantity | 1 g |
Beilstein | 2616409 |
Solubility Information | Insoluble in water. |
Formula Weight | 176.22 |
Percent Purity | 99% |
Chemical Name or Material | 2-Indanylacetic acid |