missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-4,6-dimethylaniline, 98%, Thermo Scientific Chemicals

Catalog Number p-7034208
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
4102-54-9
C8H10IN
247.079
MFCD07779005
ODPOIEACUVQCBZ-UHFFFAOYSA-N
2,4-dimethyl-6-iodoaniline, 2-iodo-4,6-di-methylaniline, benzenamine,2-iodo-4,6-dimethyl
14040290
2-iodo-4,6-dimethylaniline
CC1=CC(=C(C(=C1)I)N)C
This item is not returnable. View return policy
4102-54-9
C8H10IN
247.079
MFCD07779005
ODPOIEACUVQCBZ-UHFFFAOYSA-N
2,4-dimethyl-6-iodoaniline, 2-iodo-4,6-di-methylaniline, benzenamine,2-iodo-4,6-dimethyl
14040290
2-iodo-4,6-dimethylaniline
CC1=CC(=C(C(=C1)I)N)C

It is employed in the synthesis of benzothiazoles through copper-catalyzed one-pot three-component reactions with use of sodium hydrosulfide as a sulfur surrogate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis of benzothiazoles through copper-catalyzed one-pot three-component reactions with use of sodium hydrosulfide as a sulfur surrogate.

Solubility
Insoluble in water.

Notes
Light sensitive. Store away from light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark.
TRUSTED_SUSTAINABILITY
Melting Point 64°C to 65°C
Quantity 1 g
UN Number UN2811
Solubility Information Insoluble in water.
Formula Weight 247.08
Percent Purity 98%
Chemical Name or Material 2-Iodo-4,6-dimethylaniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'