missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Iodo-6-methoxy-4-(2-thiazolidinyl)phenol, 97%

Brand:  Alfa Aesar™ H62275.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15498818

  • 2132.99 SEK / 1g


Estimated Shipment date
from Supplier 18-02-2019
Add to basket

Specifications

Chemical Identifiers

MFCD26960673
2-(4-Hydroxy-3-iodo-5-methoxyphenyl)thiazolidine

Specifications

2-Iodo-6-methoxy-4-(2-thiazolidinyl)phenol
1g
2-(4-Hydroxy-3-iodo-5-methoxyphenyl)thiazolidine
Solid
Light Sensitive
MFCD26960673
337.18
97%
Videos
SDS