missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Iodobenzoyl chloride, 98%, Thermo Scientific Chemicals
598.81 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
250 g
Description
2-Iodobenzoyl chloride has been used in palladium-catalyzed synthesis of carbomethoxy functional group-induced isoindolin-1-ones, preparation of starting materials required for the synthesis of novel cyclic derivatives of pentavalent iodine, benziodazole oxides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Iodobenzoyl chloride has been used in palladium-catalyzed synthesis of carbomethoxy functional group-induced isoindolin-1-ones, preparation of starting materials required for the synthesis of novel cyclic derivatives of pentavalent iodine, benziodazole oxides.
Solubility
Reacts with water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
2-Iodobenzoyl chloride has been used in palladium-catalyzed synthesis of carbomethoxy functional group-induced isoindolin-1-ones, preparation of starting materials required for the synthesis of novel cyclic derivatives of pentavalent iodine, benziodazole oxides.
Solubility
Reacts with water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | 27°C to 32°C |
Boiling Point | 105°C to 106°C (1 mmHg) |
Flash Point | >110°C (230°F) |
Quantity | 10 g |
UN Number | UN3261 |
Beilstein | 2042672 |
Sensitivity | Moisture sensitive |
Synonym | o-iodobenzoyl chloride, benzoyl chloride, 2-iodo, iodobenzoyl chloride, o-iodobenzoylchloride, 2-iodobezoyl chloride, o-iodo-benzoylchloride, 2-iodobenzoic chloride, 2-iodo-benzoyl chloride, acmc-1atjg, benzoyl chloride,2-iodo |
Solubility Information | Reacts with water. |
Formula Weight | 266.47 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'