missing translation for 'onlineSavingsMsg'
Learn More

2-Iodobiphenyl, 98%, Thermo Scientific Chemicals

Catalog Number p-7044535
356.61 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
2113-51-1
C12H9I
280.108
MFCD00039396
QFUYDAGNUJWBSM-UHFFFAOYSA-N
2-iodobiphenyl, 2-iodo-1,1'-biphenyl, o-iodobiphenyl, 1,1'-biphenyl, 2-iodo, o-phenyliodobenzene, 2-iodo-biphenyl, biphenyl, 2-iodo, 2-iodo-1-phenylbenzene, 1-iodo-2-phenyl-benzene, 1,1'-biphenyl, iodo
75025
1-iodo-2-phenylbenzene
C1=CC=C(C=C1)C2=CC=CC=C2I
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
2113-51-1
C12H9I
280.108
MFCD00039396
QFUYDAGNUJWBSM-UHFFFAOYSA-N
2-iodobiphenyl, 2-iodo-1,1'-biphenyl, o-iodobiphenyl, 1,1'-biphenyl, 2-iodo, o-phenyliodobenzene, 2-iodo-biphenyl, biphenyl, 2-iodo, 2-iodo-1-phenylbenzene, 1-iodo-2-phenyl-benzene, 1,1'-biphenyl, iodo
75025
1-iodo-2-phenylbenzene
C1=CC=C(C=C1)C2=CC=CC=C2I

2-Iodobiphenyl is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodobiphenyl is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.59
Boiling Point 300°C
Flash Point >110°C (230°F)
Refractive Index 1.661
Quantity 1 g
Beilstein 2206820
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 280.11
Percent Purity 98%
Chemical Name or Material 2-Iodobiphenyl
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'