Learn More
2-Iodoethanol, 99%, stabilized, Thermo Scientific Chemicals
Catalog Number p-4510613
Click to view available options
Quantity:
10 mL
50 mL
250 mL
624-76-0
C2H5IO
171.97
MFCD00002830
QSECPQCFCWVBKM-UHFFFAOYSA-N
ethanol, 2-iodo, iodoethanol, ethylene iodohydrin, 2-iodoethan-1-ol, iodoethyl alcohol, 2-indoethanol, 2-iodo ethanol, 2-iodo-ethanol, 2-iodo-1-ethanol, 2-iodoethyl alcohol
12225
OCCI
624-76-0
C2H5IO
171.97
MFCD00002830
QSECPQCFCWVBKM-UHFFFAOYSA-N
ethanol, 2-iodo, iodoethanol, ethylene iodohydrin, 2-iodoethan-1-ol, iodoethyl alcohol, 2-indoethanol, 2-iodo ethanol, 2-iodo-ethanol, 2-iodo-1-ethanol, 2-iodoethyl alcohol
12225
OCCI
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Density 2.2050g/mL
Boiling Point 172.0°C to 175.0°C
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula ICH2CH2OH
Refractive Index 1.5690 to 1.5720
Quantity 10 mL
Beilstein 01,339
Fieser 13,348
Specific Gravity 2.205
IUPAC Name 2-iodoethanol
Stabilizer Copper
Formula Weight 171.97
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Iodoethanol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Iodoethanol, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
Hazard Statement
  • H300-Fatal if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.