Läs mer
2-Isobutyl-4,5-dimethylthiazole, 99%, Thermo Scientific™
Produktkod. 11415127
Change view
Kvantitet:
2.5 g
10 g
Förpackningsstorlek:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11415127 2.5 g 2.5g
11425127 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 11415127 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer B22554.88
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 53498-32-1
Molekylformel C9H15NS
Molekylvikt (g/mol) 169.29
MDL-nummer MFCD00053136
InChI-nyckel NSVPHVLZAKJSGV-UHFFFAOYSA-N
Synonym 2-isobutyl-4,5-dimethylthiazole, geranium thiazole, 4,5-dimethyl-2-isobutylthiazole, thiazole, 4,5-dimethyl-2-2-methylpropyl, 4,5-dimethyl-2-isobutyl thiazole, unii-5p82vd25zi, 4,5-dimethyl-2-2-methylpropyl-1,3-thiazole, acmc-1ak7t, 2-iso-butyl-4,5-dimethylthiazole, 2-isobutyl-4,5-dimethyl-thiazole
PubChem CID 104513
IUPAC-namn 4,5-dimethyl-2-(2-methylpropyl)-1,3-thiazole
LEDER CC(C)CC1=NC(C)=C(C)S1
Specifikationer
Kemiskt namn eller material 2-Isobutyl-4,5-dimethylthiazole
Densitet 0.967
Kvantitet 2.5 g
Formel vikt 169.28
Procent renhet 99%