Learn More
2-Isopropylthio-5-trifluoromethylpyridine-3-boronic acid, TRC
Product Code. 30402465
Change view
Click to view available options
Kvantitet:
100 mg
Product Code. Kvantitet
30402465 100 mg
1 options
Product Code. 30402465 Supplier Toronto Research Chemicals Supplier No. TRCI874425100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material 2-Isopropylthio-5-trifluoromethylpyridine-3-boronic acid
CAS 1256345-52-4
Molekylformel C9H11BF3NO2S
Kvantitet 100 mg
InChI formel InChI=1S/C9H11BF3NO2S/c1-5(2)17-8-7(10(15)16)3-6(4-14-8)9(11,12)13/h3-5,15-16H,1-2H3
LEDER B(C1=CC(=CN=C1SC(C)C)C(F)(F)F)(O)O
IUPAC-namn [2-propan-2-ylsulfanyl-5-(trifluoromethyl)pyridin-3-yl]boronic acid
Molekylvikt (g/mol) 265.06
Formel vikt 265.0556