Läs mer
2-Mercapto-L-histidine S-Carboxylic Acid Ethyl Ester Dihydrochloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | 2-Mercapto-L-histidine S-Carboxylic Acid Ethyl Ester Dihydrochloride |
| CAS | 909272-14-6 |
| Molekylformel | C9 H13 N3 O4 S |
| Kvantitet | 100 mg |
| Synonym | 2-Thiocarbonic Acid Ethyl Ester L-Histidine Dihydrochloride, Thiolcarbonic Acid O-Ethyl Ester 2-Mercaptohistidine Dihydrochloride |
| InChI formel | InChI=1S/C9H13N3O4S/c1-2-16-9(15)17-8-11-4-5(12-8)3-6(10)7(13)14/h4,6H,2-3,10H2,1H3,(H,11,12)(H,13,14)/t6-/m0/s1 |
| LEDER | CCOC(=O)Sc1nc(C[C@H](N)C(=O)O)c[nH]1 |
| IUPAC-namn | (2S)-2-amino-3-(2-ethoxycarbonylsulfanyl-1H-imidazol-4-yl)propanoic acid |
| Molekylvikt (g/mol) | 259.282 |
| Formel vikt | 259.063 |