Läs mer
2-Mercapto-succinic Acid diethyl ester, TRC
Produktkod. 30483098
Change view
Kvantitet:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30483098 250 mg
30477270 25 mg
30467511 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30483098 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM726125250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 23060-14-2
Molekylformel C8 H14 O4 S
Molekylvikt (g/mol) 206.26
Synonym 2-Mercaptosuccinic acid-diethyl ester, Butanedioic acid, 2-mercapto-, 1,4-diethyl ester (ACI), 1,4-Diethyl 2-mercaptobutanedioate (ACI), Butanedioic acid, mercapto-, diethyl ester (9CI), Succinic acid, mercapto-, diethyl ester (6CI, 8CI), 1,4-Diethyl 2-sulfanylbutanedioate, 2-Mercaptosuccinic acid diethyl ester, Diethyl 2-mercaptosuccinate, Diethyl 2-thiosuccinate, Diethyl mercaptosuccinate, Ethyl mercaptosuccinate
IUPAC-namn diethyl 2-sulfanylbutanedioate
LEDER CCOC(=O)CC(S)C(=O)OCC
Specifikationer
Kemiskt namn eller material 2-Mercaptosuccinic acid diethyl ester
Kvantitet 250 mg
InChI formel InChI=1S/C8H14O4S/c1-3-11-7(9)5-6(13)8(10)12-4-2/h6,13H,3-5H2,1-2H3
Formel vikt 206.0613