Learn More
2-Mercaptoethanesulfonic acid sodium salt, 98%, analytical standard, Thermo Scientific Chemicals
Catalog Number p-7077295
355.55 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
10 g
25 g
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
19767-45-4
C2H5NaO3S2
164.18
XOGTZOOQQBDUSI-UHFFFAOYSA-M
mesna, sodium 2-mercaptoethanesulfonate, mesnex, uromitexan, mitexan, 2-mercaptoethanesulfonic acid sodium salt, mistabron, mesnum, mistabronco, mucofluid
23662354
sodium;2-sulfanylethanesulfonate
C(CS(=O)(=O)[O-])S.[Na+]
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
19767-45-4
C2H5NaO3S2
164.18
XOGTZOOQQBDUSI-UHFFFAOYSA-M
mesna, sodium 2-mercaptoethanesulfonate, mesnex, uromitexan, mitexan, 2-mercaptoethanesulfonic acid sodium salt, mistabron, mesnum, mistabronco, mucofluid
23662354
sodium;2-sulfanylethanesulfonate
C(CS(=O)(=O)[O-])S.[Na+]
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Loss on Drying 2% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102% min. (On dried substance) (Iodometry)
Quantity 5 g
Merck Index 15,5979
Solubility Information Solubility in water: soluble.
Formula Weight 164.18
Percent Purity 98%
Chemical Name or Material 2-Mercaptoethanesulfonic acid sodium salt
compliance-icons
Product Identifier
  • 2-Mercaptoethanesulfonic acid sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.