Läs mer
2-Methoxy-3-methylnaphthalene, TRC
Produktkod. 30476931
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30476931 500 mg
30469290 100 mg
30475216 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30476931 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM341665500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 61873-80-1
Molekylformel C12H12O
Molekylvikt (g/mol) 172.22
IUPAC-namn 2-methoxy-3-methylnaphthalene
LEDER COc1cc2ccccc2cc1C
Specifikationer
Kemiskt namn eller material 2-Methoxy-3-methylnaphthalene
Kvantitet 500 mg
InChI formel InChI=1S/C12H12O/c1-9-7-10-5-3-4-6-11(10)8-12(9)13-2/h3-8H,1-2H3
Formel vikt 172.0888