Läs mer
2-Methoxy-3-methylphenol, TRC
Produktkod. 30467640
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30467640 25 mg
30472534 10 mg
30482772 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30467640 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM33682525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 18102-31-3
Molekylformel C8H10O2
Molekylvikt (g/mol) 138.16
Synonym 2-Methoxy-3-methylphenol, 3-Methylguaiacol
IUPAC-namn 2-methoxy-3-methylphenol
LEDER COc1c(C)cccc1O
Specifikationer
Kemiskt namn eller material 2-Methoxy-3-methylphenol
Kvantitet 25 mg
InChI formel InChI=1S/C8H10O2/c1-6-4-3-5-7(9)8(6)10-2/h3-5,9H,1-2H3
Formel vikt 138.0681