missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-3-nitropyridine-5-boronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number p-7036613
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1083168-94-8
C12H17BN2O5
280.087
MFCD12923397
XWXOPFXUTOQOSM-UHFFFAOYSA-N
2-methoxy-3-nitro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-methoxy-3-nitro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine, 2-methoxy-3-nitropyridine-5-boronic acid pinacol ester, 2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3-nitropyridine, 6-methoxy-5-nitropyridine-3-boronic acid pinacol ester, 2-methoxy-3-nitro-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-methoxy-3-nitro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, acmc-2098zq, 2-methoxy-3-nitropyridin-5-ylboronic acid pinacol ester, 6-methoxy-5-nitropyridin-3-yl boronic acid pinacol ester
57416495
2-methoxy-3-nitro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(N=C2)OC)[N+](=O)[O-]
This item is not returnable. View return policy
1083168-94-8
C12H17BN2O5
280.087
MFCD12923397
XWXOPFXUTOQOSM-UHFFFAOYSA-N
2-methoxy-3-nitro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-methoxy-3-nitro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine, 2-methoxy-3-nitropyridine-5-boronic acid pinacol ester, 2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3-nitropyridine, 6-methoxy-5-nitropyridine-3-boronic acid pinacol ester, 2-methoxy-3-nitro-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-methoxy-3-nitro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, acmc-2098zq, 2-methoxy-3-nitropyridin-5-ylboronic acid pinacol ester, 6-methoxy-5-nitropyridin-3-yl boronic acid pinacol ester
57416495
2-methoxy-3-nitro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(N=C2)OC)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 118°C to 119°C
Quantity 250 mg
Formula Weight 280.08
Percent Purity 96%
Chemical Name or Material 2-Methoxy-3-nitropyridine-5-boronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'