Läs mer
2-Methoxy-4-amino-5-ethylsulfonylbenzoic Acid, TRC
Produktkod. 17989994
Change view
Kvantitet:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17989994 25 mg
17959544 50 mg
17987444 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17989994 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM26088025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 71675-87-1
Molekylformel C10H13NO5S
Molekylvikt (g/mol) 259.28
Synonym 4-Amino-5-(ethylsulphonyl)-2-methoxybenzoic Acid, Amisulpride Imp. E (EP)
LEDER CCS(=O)(=O)c1cc(C(=O)O)c(OC)cc1N
Specifikationer
Kemiskt namn eller material 2-Methoxy-4-amino-5-ethylsulfonylbenzoic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C10H13NO5S/c1-3-17(14,15)9-4-6(10(12)13)8(16-2)5-7(9)11/h4-5H,3,11H2,1-2H3,(H,12,13)
Procent renhet >95%