missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-pyridinecarboxylic acid, 97%, Thermo Scientific™

Catalog Number 11986431
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
105596-63-2
C7H7NO3
153.14
DPNDWFVORIGXQO-UHFFFAOYSA-N
2-methoxyisonicotinic acid, 2-methoxy-4-pyridinecarboxylic acid, 2-methoxy-isonicotinic acid, 4-pyridinecarboxylic acid, 2-methoxy, 2-methoxy-isonicotinicacid, 2-methoxy-4-picolinic acid, pubchem9293, ksc179m4j, 4-pyridinecarboxylicacid, 2-methoxy, 2-methyloxy-4-pyridinecarboxylic acid
819942
2-methoxypyridine-4-carboxylic acid
COC1=NC=CC(=C1)C(=O)O
This item is not returnable. View return policy
105596-63-2
C7H7NO3
153.14
DPNDWFVORIGXQO-UHFFFAOYSA-N
2-methoxyisonicotinic acid, 2-methoxy-4-pyridinecarboxylic acid, 2-methoxy-isonicotinic acid, 4-pyridinecarboxylic acid, 2-methoxy, 2-methoxy-isonicotinicacid, 2-methoxy-4-picolinic acid, pubchem9293, ksc179m4j, 4-pyridinecarboxylicacid, 2-methoxy, 2-methyloxy-4-pyridinecarboxylic acid
819942
2-methoxypyridine-4-carboxylic acid
COC1=NC=CC(=C1)C(=O)O
CAS Min % 96.0
CAS Max % 100.0
Melting Point 186°C to 188°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 153.14
Percent Purity 97%
Physical Form Crystalline Powder or Solid
Chemical Name or Material 2-Methoxy-4-pyridinecarboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methoxy-4-pyridinecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'