missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxy-5-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Mostly existing as pharmaceutical and agrochemical products.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
Mostly existing as pharmaceutical and agrochemical products.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
Specifications
Melting Point | 34°C to 37°C |
Quantity | 1 g |
UN Number | UN3261 |
Solubility Information | Insoluble in water. |
Formula Weight | 269.06 |
Percent Purity | 97% |
Chemical Name or Material | 2-Methoxy-5-(trifluoromethyl)benzyl bromide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'