missing translation for 'onlineSavingsMsg'
Läs mer

2-Methoxyethyl Acetate, TRC

Product Code. 30133235
Change view
Click to view available options
Kvantitet:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30133235 25 g
30133587 5 g
30005630 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133235 Supplier Toronto Research Chemicals Supplier No. TRCM30448025G

Please to purchase this item. Need a web account? Register with us today!

Denna vara kräver en prisuppdatering innan den beställs.

Begär ett pris för denna vara här.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-49-6
Molekylformel C5 H10 O3
Molekylvikt (g/mol) 118.13
Synonym Ethanol, 2-methoxy-, acetate (6CI,7CI,8CI,9CI), 1-Acetoxy-2-methoxyethane, 2-Methoxyethanol acetate, 2-Methoxyethyl acetate, Acetyl methyl cellosolve, Ethylene glycol acetate monomethyl ether, Ethylene glycol methyl ether acetate, Ethylene glycol monomethyl ether acetate, Glycol monomethyl ether acetate, MC acetate, Methyl Cellosolve acetate, Methyl glycol acetate, β-Methoxyethyl acetate
IUPAC-namn 2-methoxyethyl acetate
LEDER COCCOC(=O)C

Specifications

Kemiskt namn eller material 2-Methoxyethyl acetate
Kvantitet 25 g
InChI formel InChI=1S/C5H10O3/c1-5(6)8-4-3-7-2/h3-4H2,1-2H3
Formel vikt 118.063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.