Läs mer
2-Methoxymethyl Methansulfonate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | 2-Methoxymethyl Methansulfonate |
| CAS | 16427-44-4 |
| Molekylformel | C4H10O4S |
| Kvantitet | 100 g |
| Synonym | 2-Methoxyethanol Methanesulfonate, Methanesulfonic Acid 2-Methoxyethyl Ester, 2-Methoxyethyl Mesylate, 2-Methoxyethyl Methanesulfonate, Ethylene Glycol Methanesulfonate Methyl Ether, Ethylene Gycol Monomethyl Ether Methanesulfonate, beta-Methoxyethyl Methanesulfonate |
| InChI formel | InChI=1S/C4H10O4S/c1-7-3-4-8-9(2,5)6/h3-4H2,1-2H3 |
| LEDER | COCCOS(=O)(=O)C |
| IUPAC-namn | 2-methoxyethyl methanesulfonate |
| Molekylvikt (g/mol) | 154.18 |
| Formel vikt | 154.03 |