missing translation for 'onlineSavingsMsg'
Learn More

2-(Methoxymethyl)phenylboronic acid, 98%, Thermo Scientific™

Catalog Number p-7077491
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
126617-98-9
C8H11BO3
165.99
JHPPAFXQJZJGEP-UHFFFAOYSA-N
2-methoxymethylphenylboronic acid, 2-methoxymethylbenzeneboronic acid, 2-methoxymethyl phenyl boronic acid, 2-methoxymethyl phenylboronic acid, 2-methoxymethyl benzeneboronic acid, 2-methoxymethylphebylboronic acid, boronic acid, 2-methoxymethyl phenyl, pubchem7997
3759408
[2-(methoxymethyl)phenyl]boronic acid
B(C1=CC=CC=C1COC)(O)O
This item is not returnable. View return policy
126617-98-9
C8H11BO3
165.99
JHPPAFXQJZJGEP-UHFFFAOYSA-N
2-methoxymethylphenylboronic acid, 2-methoxymethylbenzeneboronic acid, 2-methoxymethyl phenyl boronic acid, 2-methoxymethyl phenylboronic acid, 2-methoxymethyl benzeneboronic acid, 2-methoxymethylphebylboronic acid, boronic acid, 2-methoxymethyl phenyl, pubchem7997
3759408
[2-(methoxymethyl)phenyl]boronic acid
B(C1=CC=CC=C1COC)(O)O
Melting Point 42.0°C to 44.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 165.99
Percent Purity 98%
Chemical Name or Material 2-(Methoxymethyl)phenylboronic acid
compliance-icons
Product Identifier
  • 2-(Methoxymethyl)phenylboronic acid, 98%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'