missing translation for 'onlineSavingsMsg'
Läs mer

2-Methoxyphenol-13C6, TRC

Produktkod. 30481110
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mg
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30481110 25 mg -
30484072 1 mg -
30465221 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30481110 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM26530325MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 202326-52-1
Molekylformel C13C6H8O2
Molekylvikt (g/mol) 130.09
Synonym o-Methoxy-phenol-13C6, 1-Hydroxy-2-methoxybenzene-13C6, 2-Hydroxyanisole-13C6, Anastil-13C6, Guaiacol-13C6, Guaiastil-13C6, Methylcatechol-13C6, o-Hydroxyanisole-13C6, O-Methoxyphenol-13C6, NSC 3815-13C6
IUPAC-namn 2-methoxyphenol-13C6
LEDER O[13C]1=[13C]([H])[13C]([H])=[13C]([H])[13C]([H])=[13C]1OC

Specifikationer

Kemiskt namn eller material 2-Methoxyphenol-13C6
Kvantitet 25 mg
InChI formel InChI=1S/C7H8O2/c1-9-7-5-3-2-4-6(7)8/h2-5,8H,1H3/i2+1,3+1,4+1,5+1,6+1,7+1
Formel vikt 130.07
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.