Learn More
2-Methoxyphenyl isocyanate, 98%, Thermo Scientific Chemicals
Catalog Number p-7023976
Click to view available options
Quantity:
5 g
25 g
700-87-8
C8H7NO2
149.15
MFCD00002004
SUVCZZADQDCIEQ-UHFFFAOYSA-N
2-methoxyphenyl isocyanate, 2-methoxyphenylisocyanate, benzene, 1-isocyanato-2-methoxy, o-methoxyphenyl isocyanate, 2-isocyanatoanisole, 1-isocyanato-2-methoxy-benzene, o-anisylisocyanate, 2-methoxybenzenisocyanate, o-anisyl isocyanate, methoxyphenyl isocyanate
69695
1-isocyanato-2-methoxybenzene
COC1=CC=CC=C1N=C=O
700-87-8
C8H7NO2
149.15
MFCD00002004
SUVCZZADQDCIEQ-UHFFFAOYSA-N
2-methoxyphenyl isocyanate, 2-methoxyphenylisocyanate, benzene, 1-isocyanato-2-methoxy, o-methoxyphenyl isocyanate, 2-isocyanatoanisole, 1-isocyanato-2-methoxy-benzene, o-anisylisocyanate, 2-methoxybenzenisocyanate, o-anisyl isocyanate, methoxyphenyl isocyanate
69695
1-isocyanato-2-methoxybenzene
COC1=CC=CC=C1N=C=O
2-Methoxyphenyl isocyanate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Methoxyphenyl isocyanate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Hydrolyzes with water.
Notes
Moisture Sensitive. Protect from water and humidity. Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store under dry inert gas.
TRUSTED_SUSTAINABILITY
Density 1.122
Boiling Point 94°C to 96°C (10 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.5414
Quantity 5 g
UN Number UN2206
Beilstein 387700
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes with water.
Formula Weight 149.15
Percent Purity 98%
Chemical Name or Material 2-Methoxyphenyl isocyanate