Läs mer
2-Methoxyphenylsulfate Potassium Salt, TRC
Produktkod. 30138735
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138735 1 g
30005030 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30138735 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2663101G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6100-07-8
Molekylformel C7 H8 O5 S . K
Molekylvikt (g/mol) 243.29
Synonym Guaiacol, hydrogen sulfate, potassium salt (6CI), Phenol, 2-methoxy-, hydrogen sulfate, potassium salt (9CI), Phenol, o-methoxy-, hydrogen sulfate, potassium salt (8CI), Guiacolsulfonic acid potassium salt, Potassium o-methoxyphenyl sulfate
IUPAC-namn 2-methoxyphenyl hydrogen sulfate, potassium salt
LEDER [K].O=S(=O)(O)OC=1C=CC=CC1OC
Specifikationer
Kemiskt namn eller material 2-Methoxyphenylsulfate Potassium Salt
Kvantitet 1 g
InChI formel InChI=1S/C7H8O5S.K/c1-11-6-4-2-3-5-7(6)12-13(8,9)10;/h2-5H,1H3,(H,8,9,10);
Formel vikt 242.97