Läs mer
2-Methoxyresorcinol, TRC
Produktkod. 30468233
Change view
Kvantitet:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30468233 5 g
30481042 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30468233 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2676005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Methoxyresorcinol
CAS 29267-67-2
Molekylformel C7 H8 O3
Kvantitet 5 g
InChI formel InChI=1S/C7H8O3/c1-10-7-5(8)3-2-4-6(7)9/h2-4,8-9H,1H3
LEDER COc1c(O)cccc1O
IUPAC-namn 2-methoxybenzene-1,3-diol
Molekylvikt (g/mol) 140.14
Formel vikt 140.0473